Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 181183-52-8 |
| Molecular Formula | C21H31N3O7S |
| Quantity | 250 mg |
| Solubility Information | Solubility aq. soln.: soluble. |
| InChI Key | QHATUKWEVNMHRY-UHFFFAOYNA-N |
| SMILES | OC(CC(O)=O)C(O)=O.CN(C)CCC1=CNC2=CC=C(CS(=O)(=O)N3CCCC3)C=C12 |
| IUPAC Name | 2-hydroxybutanedioic acid; dimethyl(2-{5-[(pyrrolidine-1-sulfonyl)methyl]-1H-indol-3-yl}ethyl)amine |
| Molecular Weight (g/mol) | 469.55 |
| Formula Weight | 469.55 |
| Chemical Name or Material | Almotriptan malate |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.