missing translation for 'onlineSavingsMsg'
Learn More

Alogliptin Benzoate, MedChemExpress

Product Code. 16448268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448268 10 mg 10mg
16437996 100 mg 100mg
16434157 50 mg 50mg
16430708 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16448268 Supplier Medchem Express Supplier No. HYA0023
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alogliptin Benzoate (SYR-322) is a potent, selective and orally active inhibitor of DPP-4 with an IC50 of <10 nM, and exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9. Alogliptin Benzoate can be used for the research of type 2 diabetes.

Alogliptin Benzoate (SYR-322) is a potent, selective and orally active inhibitor of DPP-4 with an IC50 of <10 nM, and exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9. Alogliptin Benzoate can be used for the research of type 2 diabetes.

Specifications

Chemical Name or Material Alogliptin Benzoate
CAS 850649-62-6
Color White
Purity Grade Notes Research
Molecular Formula C25H27N5O4
Quantity 10 mg
Synonym SYR 322
Solubility Information DMSO : 50 mg/mL (108.34 mM; Need ultrasonic)
SMILES O=C(C=C(N1C[C@H](N)CCC1)N(C2=O)CC3=C(C#N)C=CC=C3)N2C.O=C(O)C4=CC=CC=C4
Molecular Weight (g/mol) 461.51
Formula Weight 461.51
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.