missing translation for 'onlineSavingsMsg'
Learn More

Alogliptin Benzoate, TRC

Product Code. 30132893
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132893 250 mg 250mg
30007103 10 mg 10mg
30003892 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132893 Supplier Toronto Research Chemicals Supplier No. TRCA575440250MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850649-62-6
Molecular Formula C18 H21 N5 O2 . C7 H6 O2
Molecular Weight (g/mol) 461.51
Synonym Benzonitrile, 2-[[6-[(3R)-3-amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-, benzoate (1:1), 2-[[6-[(3R)-3-Amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]benzonitrile Benzoate (1:1), Benzonitrile, 2-[[6-[(3R)-3-amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-, monobenzoate (9CI), Alogliptin benzoate, Nesina, SYR 322
IUPAC Name 2-[[6-[(3R)-3-aminopiperidin-1-yl]-3-methyl-2,4-dioxopyrimidin-1-yl]methyl]benzonitrile;benzoic acid
SMILES CN1C(=O)C=C(N2CCC[C@@H](N)C2)N(Cc3ccccc3C#N)C1=O.OC(=O)c4ccccc4

Specifications

InChI Formula InChI=1S/C18H21N5O2.C7H6O2/c1-21-17(24)9-16(22-8-4-7-15(20)12-22)23(18(21)25)11-14-6-3-2-5-13(14)10-19;8-7(9)6-4-2-1-3-5-6/h2-3,5-6,9,15H,4,7-8,11-12,20H2,1H3;1-5H,(H,8,9)/t15-;/m1./s1
Formula Weight 461.2063
Percent Purity >95%
Quantity 250 mg
Chemical Name or Material Alogliptin Benzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.