missing translation for 'onlineSavingsMsg'
Learn More

Alpaca anti-Mouse IgG2b Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 488, Invitrogen™

Product Code. 17172234
Change view
Click to view available options
Quantity:
10 μL
Unit Size:
10µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17172234 10 μL 10µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17172234 Supplier Invitrogen Supplier No. SA510334

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alpaca Recombinant Monoclonal Antibody

Biclonal nanobodies specific for mouse IgG2b Fc. CTK0105: KD = 5 nM, CTK0106: KD = 0.2 nM Non Cross-Reactivity includes: Rabbit, rat, goat, sheep, guinea pig, human, and macaque serum and to mouse IgG1, IgG2a, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG2b primary antibody and Alpaca anti-mouse IgG2b Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek

Mouse IgG2b Nano (VHH) Recombinant Monoclonal antibody specifically detects Mouse IgG2b Nano (VHH) in Mouse samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2b Nano (VHH)
Applications Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
Classification Monoclonal
Clone CTK0105/CTK0106
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7
Gene Alias Biclonal; Fc-specific recombinant VHH; Nanobodies
Host Species Alpaca
Immunogen Purified mouse IgG2b
Purification Method Affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.