Learn More
Alpaca anti-Mouse IgG2b Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Description
Biclonal nanobodies specific for mouse IgG2b Fc. CTK0105: KD = 5 nM, CTK0106: KD = 0.2 nM Non Cross-Reactivity includes: Rabbit, rat, goat, sheep, guinea pig, human, and macaque serum and to mouse IgG1, IgG2a, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG2b primary antibody and Alpaca anti-mouse IgG2b Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek
Specifications
Specifications
| Antigen | Mouse IgG2b Nano (VHH) |
| Applications | Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot |
| Classification | Monoclonal |
| Clone | CTK0105/CTK0106 |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7 |
| Gene Alias | Biclonal; Fc-specific recombinant VHH; Nanobodies |
| Host Species | Alpaca |
| Immunogen | Purified mouse IgG2b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.