missing translation for 'onlineSavingsMsg'
Learn More

Alpaca monoclonal [10A4] Anti-Mouse IgG1 antibody (Alexa Fluor™ 647), Abcam

Product Code. 32114608 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32114608 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32114608 Supplier Abcam Supplier No. ab325287100uL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alpaca Monoclonal Antibody

Recombinant secondary antibody suitable for ICC/IF. Ideal for fluorescent cell and tissue imaging.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ighg1
Applications Immunocytochemistry/Immunofluorescence
Cellular Localization Isoform Secreted: Secreted
Classification Monoclonal
Clone '10A4
Conjugate Alexa Fluor 647
Formulation Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene Ighg1
Gene Accession No. P01868
Host Species Alpaca
Immunogen Ighg1
Purification Method Size-exclusion chromatography
Quantity 100 μL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -80°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene Ighg1
Target Molecule Ighg1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.