missing translation for 'onlineSavingsMsg'
Learn More

Alpaca multiclonal [10E10 / 8C10] Anti-Rabbit IgG antibody (ATTO 488), Abcam

Product Code. 32120322 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120322 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120322 Supplier Abcam Supplier No. ab325297100uL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alpaca Polyclonal Antibody

Recombinant secondary antibody suitable for ICC/IF.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Rabbit IgG Fc (IGHG)
Applications Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Clone '10E10 / 8C10
Conjugate Atto 488
Formulation Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene Rabbit IgG Fc
Gene Accession No. P01870
Host Species Alpaca
Immunogen Rabbit IgG Fc (IGHG)
Purification Method Size-exclusion chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage -80°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene Rabbit IgG Fc
Target Molecule Rabbit IgG Fc (IGHG)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.