missing translation for 'onlineSavingsMsg'
Learn More

alpha-[2-(Methylamino)ethyl]benzyl Alcohol, TRC

Product Code. 17993885
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993885 5 g 5g
17931635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17993885 Supplier Toronto Research Chemicals Supplier No. TRCM2861205G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42142-52-9
Molecular Formula C10 H15 N O
Molecular Weight (g/mol) 165.23
Synonym (1RS)-3-(Methylamino)-1-phenylpropan-1-ol, Fluoxetine Hydrochloride Imp. A (EP), Benzenemethanol, α-[2-(methylamino)ethyl]-, Benzyl alcohol, α-[2-(methylamino)ethyl]- (6CI,7CI), α-[2-(Methylamino)ethyl]benzenemethanol, (±)-N-Methyl-3-hydroxy-3-phenylpropylamine, 3-(Methylamino)-1-phenyl-1-propanol, 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-Methylamino-1-phenylpropanol, 3-N-Methylamino-1-phenyl-1-propanol, Methyl(3-hydroxy-3-phenylpropyl)amine, N-(3-Hydroxy-3-phenylpropyl)-N-methylamine, N-(3-Hydroxy-3-phenylpropyl)methylamine, N-Methyl-3-hydroxy-3-phenylpropylamine, N-Methyl-3-phenyl-3-hydroxypropylamine, N-Methyl-N-(3-hydroxy-3-phenylpropyl)amine
IUPAC Name 3-(methylamino)-1-phenylpropan-1-ol
SMILES CNCCC(O)c1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H15NO/c1-11-8-7-10(12)9-5-3-2-4-6-9/h2-6,10-12H,7-8H2,1H3
Formula Weight 165.1154
Chemical Name or Material (1RS)-3-(Methylamino)-1-phenylpropan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.