missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Acetamidocinnamic Acid, TRC

Product Code. 30317799
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317799 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317799 Supplier Toronto Research Chemicals Supplier No. TRCA150000100G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material a-Acetamidocinnamic Acid
CAS 5469-45-4
Molecular Formula C11 H11 N O3
Quantity 100 g
Synonym 2-Propenoic acid, 2-(acetylamino)-3-phenyl-, Cinnamic acid, α-acetamido- (6CI,7CI,8CI), 2-(Acetylamino)-3-phenyl-2-propenoic acid, 2-Acetamidocinnamic acid, 2-Acetylamino-3-phenylacrylic acid, 2-Acetylamino-3-phenylprop-2-enoic acid, N-Acetyldehydrophenylalanine, NSC 25333, α-Acetamidocinnamic acid, α-Acetylaminocinnamic acid
InChI Formula InChI=1S/C11H11NO3/c1-8(13)12-10(11(14)15)7-9-5-3-2-4-6-9/h2-7H,1H3,(H,12,13)(H,14,15)/b10-7-
SMILES CC(=O)N\C(=C/c1ccccc1)\C(=O)O
IUPAC Name (Z)-2-acetamido-3-phenylprop-2-enoic acid
Molecular Weight (g/mol) 205.21
Formula Weight 205.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.