missing translation for 'onlineSavingsMsg'
Learn More

alpha-Acetyl-gamma-butyrolactone, 98+%

Product Code. 11496116
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496116 500 g 500g
11486116 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496116 Supplier Thermo Scientific Alfa Aesar Supplier No. B22967.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 517-23-7 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-23-7
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005394
InChI Key OMQHDIHZSDEIFH-UHFFFAOYNA-N
Synonym 2-acetylbutyrolactone, 3-acetyldihydrofuran-2 3h-one, alpha-acetylbutyrolactone, alpha-acetyl-gamma-butyrolactone, 3-acetyldihydro-2 3h-furanone, 2 3h-furanone, 3-acetyldihydro, 3-acetyltetrahydrofuran-2-one, 2-oxo-3-acetyltetrahydrofuran, 3-acetyltetrahydro-2-furanone, 3-acetyl-2 3h-4,5-dihydrofuranone
PubChem CID 10601
IUPAC Name 3-acetyloxolan-2-one
SMILES CC(=O)C1CCOC1=O

Specifications

Density 1.19
Boiling Point 107°C to 108°C (5 mmHg)
Flash Point 135°C (275°F)
Refractive Index 1.459
Quantity 500 g
Beilstein 112676
Merck Index 14,83
Formula Weight 128.13
Percent Purity ≥98%
Chemical Name or Material alpha-Acetyl-gamma-butyrolactone
compliance-icons
Product Identifier
  • alpha-Acetyl-gamma-Butyrolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: alpha-Acetyl-gamma-Butyrolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.