missing translation for 'onlineSavingsMsg'
Learn More

Alpha,Alpha-Bis[2-(dimethylamino)ethyl]-4-chlorophenylacetonitrile, TRC

Product Code. 30003604
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003604 2.5 g 2.5g
18062400 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003604 Supplier Toronto Research Chemicals Supplier No. TRCB4298002.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246816-57-8
Molecular Formula C16 H24 Cl N3
Molecular Weight (g/mol) 293.83
Synonym Benzeneacetonitrile, 4-chloro-α,α-bis[2-(dimethylamino)ethyl]-, 2-(4-Chlorophenyl)-4-(dimethylamino)-2-[2-(dimethylamino)ethyl]butanenitrile, Chlorphenamine Maleate Imp. A (EP), 4-chloro-alpha,alpha-bis[2-(dimethylamino)ethyl]-;
IUPAC Name 2-(4-chlorophenyl)-4-(dimethylamino)-2-[2-(dimethylamino)ethyl]butanenitrile
SMILES CN(C)CCC(CCN(C)C)(C#N)c1ccc(Cl)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C16H24ClN3/c1-19(2)11-9-16(13-18,10-12-20(3)4)14-5-7-15(17)8-6-14/h5-8H,9-12H2,1-4H3
Formula Weight 293.1659
Percent Purity >95%
Chemical Name or Material 2-(4-Chlorophenyl)-4-(dimethylamino)-2-[2-(dimethylamino)ethyl]butanenitrile
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.