missing translation for 'onlineSavingsMsg'
Learn More

alpha-Aminocyclohexanecarboxylic Acid, TRC

Product Code. 30318466
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30318466 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30318466

Brand: Toronto Research Chemicals TRCA6042201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Aminocyclohexanecarboxylic Acid
CAS 2756-85-6
Molecular Formula C7 H13 N O2
Quantity 1 g
Synonym 1-Aminocyclohexane-1-carboxylic Acid, 1-Aminocyclohexanecarboxylic Acid, Homocycloleucine, NSC 9059
InChI Formula InChI=1S/C7H13NO2/c8-7(6(9)10)4-2-1-3-5-7/h1-5,8H2,(H,9,10)
SMILES NC1(CCCCC1)C(=O)O
IUPAC Name 1-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 143.184
Formula Weight 143.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.