missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Amylcinnamaldehyde stabilized with BHT, TRC
Specifications
Specifications
| Chemical Name or Material | Amylcinnamic aldehyde |
| CAS | 122-40-7 |
| Molecular Formula | C14 H18 O |
| Quantity | 5 mL |
| Synonym | Cinnamaldehyde, α-amyl- (4CI), Cinnamaldehyde, α-pentyl- (6CI,7CI,8CI), 2-(Phenylmethylene)heptanal, 2-Benzylideneheptanal, Amyl cinnamal, Amylcinnamaldehyde, Amylcinnamic acid aldehyde, Amylcinnamic aldehyde, Flomine, Jasminal, Jasminaldehyde, Jasmine aldehyde, NSC 6649, Pentylcinnamaldehyde, α-Amyl-β-phenylacrolein, α-Amylcinnamal, α-Amylcinnamaldehyde, α-Pentylcinnamaldehyde |
| InChI Formula | InChI=1S/C14H18O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11-12H,2-3,5,10H2,1H3/b14-11+ |
| SMILES | CCCCC\C(=C/c1ccccc1)\C=O |
| IUPAC Name | (2E)-2-benzylideneheptanal |
| Molecular Weight (g/mol) | 202.29 |
| Formula Weight | 202.1358 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?