missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bisabolol, 96%

Product Code. 11406974
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406974 10 g 10g
11416974 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406974 Supplier Thermo Scientific Alfa Aesar Supplier No. B21457.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 515-69-5 | C15H26O | 222.372 g/mol

It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

Solubility
Soluble in alcohol, natural, mineral and synthetic oils. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 515-69-5
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD03846910
InChI Key RGZSQWQPBWRIAQ-LSDHHAIUSA-N
Synonym bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260
PubChem CID 1549992
IUPAC Name (2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
SMILES CC1=CCC(CC1)C(C)(CCC=C(C)C)O

Specifications

Density 0.93
Flash Point 135°C (275°F)
Refractive Index 1.495
Quantity 10 g
Beilstein 5733954
Sensitivity Air Sensitive
Merck Index 14,1241
Solubility Information Soluble in alcohol,natural,mineral and synthetic oils. Insoluble in water.
Formula Weight 222.37
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bisabolol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.