missing translation for 'onlineSavingsMsg'
Learn More

alpha-Boswellic acid, MedChemExpress

Product Code. 16479038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479038 10 mg 10mg
16451458 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479038 Supplier Medchem Express Supplier No. HYN0611
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress alpha-Boswellic acid (α-Boswellic acid) is a pentacyclic triterpene compound from extracts of Frankincense, has anticonvulsant and anti-cancer properties. alpha-Boswellic acid prevents and decreases the progression of Alzheimer’s hallmarks in vivo and can be used for Alzheimer’s disease research.

alpha-Boswellic acid (α-Boswellic acid) is a pentacyclic triterpene compound from extracts of Frankincense, has anticonvulsant and anti-cancer properties. alpha-Boswellic acid prevents and decreases the progression of Alzheimer’s hallmarks in vivo and can be used for Alzheimer’s disease research.

Specifications

Chemical Name or Material alpha-Boswellic acid
CAS 471-66-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H48O3
Quantity 10 mg
Synonym α-Boswellic acid
Solubility Information DMSO : 25 mg/mL (54.74 mM; Need ultrasonic)
SMILES C[C@@]([C@@]12C)(CC[C@@]3([H])[C@@]4(C)C(O)=O)[C@@](CC=C1[C@@](CC(C)(C)CC5)([H])[C@@]5(C)CC2)([H])[C@]3(CC[C@H]4O)C
Molecular Weight (g/mol) 456.7
Formula Weight 456.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.