missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-gamma-butyrolactone, 97%

Product Code. 11433587
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433587 100 g 100g
11423587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433587 Supplier Thermo Scientific Alfa Aesar Supplier No. A17344.22
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5061-21-2 | C4H5BrO2 | 164.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5061-21-2
Molecular Formula C4H5BrO2
Molecular Weight (g/mol) 164.99
MDL Number MFCD00005387
InChI Key LFJJGHGXHXXDFT-UHFFFAOYNA-N
Synonym 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone
PubChem CID 95463
IUPAC Name 3-bromooxolan-2-one
SMILES BrC1CCOC1=O

Specifications

Density 1.994
Boiling Point 137°C to 139°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.508
Quantity 100 g
Beilstein 107800
Sensitivity Moisture sensitive
Formula Weight 164.99
Percent Purity 97%
Chemical Name or Material alpha-Bromo-gamma-butyrolactone
compliance-icons
Product Identifier
  • alpha-Bromo-gamma-butyrolactone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: alpha-Bromo-gamma-butyrolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.