missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Cellulose (powdered), TRC

Product Code. 30135284
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135284 500 g 500g
17997694 50 g 50g
17999734 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135284 Supplier Toronto Research Chemicals Supplier No. TRCC459200500G
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6
Molecular Formula (C6H10O5)n
Molecular Weight (g/mol) 504.44
Synonym Cellulose, β-Amylose
IUPAC Name (2R,3R,4R,5S,6R)-6-(hydroxymethyl)-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triol
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@@H]2[C@@H](CO)O[C@@H](O[H])[C@H](O)[C@H]2O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11-,12+/m1/s1
Formula Weight 504.169
Chemical Name or Material Silicified Microcystalline Cellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.