missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Chloralose, 98+%, beta anomer ca 20%

Product Code. 11480393
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480393 25 g 25g
11490393 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480393 Supplier Thermo Scientific Chemicals Supplier No. A11492.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15879-93-3 | C8H11Cl3O6 | 309.52 g/mol

It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canadensis) and blue geese (Chen caerulescens) were caught with alpha-chloralose, an oral hypnotic administered on bait. Wild turkeys (Meleagris gallopavo) were captured with alpha-chloralose, an oral anesthetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canaden
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15879-93-3
Molecular Formula C8H11Cl3O6
Molecular Weight (g/mol) 309.52
MDL Number MFCD00005542
InChI Key OJYGBLRPYBAHRT-OPKHMCHVSA-N
Synonym a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol
PubChem CID 7057995
SMILES OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

Specifications

Melting Point 180°C to 184°C
Quantity 25 g
Beilstein 85418
Merck Index 14,2072
Solubility Information Slightly soluble in water.
IUPAC Name 1-[(2R,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
Formula Weight 309.53
Percent Purity ≥98%
Chemical Name or Material alpha-Chloralose, beta anomer ≈ 15%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.