missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin, 97+%

Product Code. 15453419
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453419 25 g 25g
15443419 5 g 5g
15463419 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15453419 Supplier Thermo Scientific Alfa Aesar Supplier No. J60687.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Useful for selective precipitation of enantiomeric, positional or structural isomers | CAS: 10016-20-3 | C36H60O30 | 972.846 g/mol

Useful for selective precipitation of enantiomeric, positional or structural isomersα-Cyclodextrin is used as a fiber ingredient, an odor or flavor masking agent. It is also useful for emulsification applications. It is also used as whipping fiber and emulsifying fiber. It finds application in medical, healthcare and food and beverage applications. It is also used to lower blood low-density lipoprotein cholesterol levels and lower blood triglyceride levels. It plays an essential role in fat free or fat containing dessert compositions and also employed for the reduction or the replacement of egg white in confectionary and bakery applications. Further, it acts as a supramolecular carrier, complexing agent and controlled drug release. In addition to this, it is used to increase the insulin and leptin sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 1%(w/v)

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
MDL Number MFCD00078207
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 25 g
Merck Index 14,2718
Solubility Information Soluble in water at 1%(w/v)
Optical Rotation +136° (c=10 in Water)
Formula Weight 972.84
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material alpha-Cyclodextrin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.