missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclohexylphenylacetonitrile, 98%

Product Code. 11327415
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327415 50 g 50g
11317415 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327415 Supplier Thermo Scientific Alfa Aesar Supplier No. L05350.18
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3893-23-0 | C14H17N | 199.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3893-23-0
Molecular Formula C14H17N
Molecular Weight (g/mol) 199.30
MDL Number MFCD00019362
InChI Key IZSWBXTYTALSOZ-UHFFFAOYNA-N
Synonym cyclohexylphenylacetonitrile, cyclohexyl phenyl acetonitrile, alpha-cyclohexylphenylacetonitrile, benzeneacetonitrile, .alpha.-cyclohexyl, 2-cyclohexyl-2-phenylethanenitrile, acmc-20alcf, cyclohexyl-phenylacetonitrile, alpha-phenylcyclohexaneacetonitrile, 2-cyclohexyl-2-phenyl-acetonitrile, .alpha.-cyclohexylphenylacetonitrile
PubChem CID 95302
IUPAC Name 2-cyclohexyl-2-phenylacetonitrile
SMILES N#CC(C1CCCCC1)C1=CC=CC=C1

Specifications

Melting Point 52°C to 56°C
Boiling Point 137°C to 140°C (3 mmHg)
Quantity 50 g
UN Number UN3439
Beilstein 2051115
Formula Weight 199.3
Percent Purity 98%
Chemical Name or Material alpha-Cyclohexylphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.