missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclopentylmandelic acid, 98%

Product Code. 10448142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10448142 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10448142 Supplier Thermo Scientific Acros Supplier No. 250970050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 427-49-6 | C13H16O3 | 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 427-49-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 144.0°C to 150.0°C
Color White to Beige
Boiling Point 110.0°C (0.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C13H16O3
MDL Number MFCD00019296
Quantity 5 g
Synonym alpha-cyclopentylmandelic acid, cyclopentylphenylglycolic acid, cyclopentyl hydroxy phenylacetic acid, cyclopentyl-hydroxy-phenyl-acetic acid, cyclopentyl mandelic acid, alpha-cyclopentyl-dl-mandelic acid, a-cyclo pentyl mandelic acid, alpha-cyclopentylmandelicacid, 2-rs-cyclopentyl-2-hydroxy-2-phenylacetic acid alpha-cyclopentylmandelic acid, cyclopentylmandelic acid
Solubility Information Solubility in water: .. Other solubilities: soluble in bases
InChI Key WFLUEQCOAQCQLP-UHFFFAOYSA-N
SMILES C1CCC(C1)C(C2=CC=CC=C2)(C(=O)O)O
IUPAC Name 2-cyclopentyl-2-hydroxy-2-phenylacetic acid
Molecular Weight (g/mol) 220.27
PubChem CID 98283
Formula Weight 220.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material α-Cyclopentylmandelic acid
Show More Show Less
compliance-icons
Product Identifier
  • alpha-Cyclopentylmandelic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.