missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Galactose 1-Phosphate Dipotassium Salt, TRC

Product Code. 30003221
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003221 100 mg 100mg
30009113 50 mg 50mg
30004987 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003221 Supplier Toronto Research Chemicals Supplier No. TRCG155760100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19046-60-7
Molecular Formula C6 H11 K2 O9 P
Molecular Weight (g/mol) 336.316
Synonym α-D-Galactopyranose 1-(Dihydrogen Phosphate) Dipotassium Salt, α-D-Galactopyranose-1 1-Phosphate Dipotassium Salt, D-Galactose 1-Phosphate Dipotassium Salt, α-D-Galactopyranosyl Phosphate Dipotassium Salt, α-D-Galactose 1-Phosphate Dipotassium Salt, α-D-Galactosyl Phosphate Dipotassium Salt
IUPAC Name [potassiooxy-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-phosphoryl]oxypotassium
SMILES [K+].[K+].OC[C@H]1O[C@H](OP(=O)([O-])[O-])[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H13O9P.2K/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;/h2-10H,1H2,(H2,11,12,13);;/q;2*+1/p-2/t2-,3+,4+,5-,6-;;/m1../s1
Formula Weight 335.9415
Chemical Name or Material alpha-D-Galactose 1-Phosphate Dipotassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.