missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Galactose pentaacetate

Product Code. 15497689
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497689 5 g 5g
15407699 25 g 25g
15417699 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497689 Supplier Thermo Scientific Alfa Aesar Supplier No. J66366.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4163-59-1 | C16H22O11 | 390.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4163-59-1
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-galactose pentaacetate, a-d-galactose pentaacetate, pentaacetyl-alpha-d-galactose, alpha-d-galactose penta-acetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactopyranose, alpha-d-galactose-pentaacetate, 1,2,3,4,6-pentaacetyl-alpha-d-galactose, alpha-d-galactose 1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactose, penta-o-acetyl-alpha-d-galactose
PubChem CID 6432426
ChEBI CHEBI:63145
IUPAC Name [(2R,3S,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 143°C to 144°C
Color White
Quantity 5 g
Solubility Information Soluble in methanol at approximately 50mg/ml
Formula Weight 390.34
Physical Form Powder
Chemical Name or Material alpha-D-Galactose pentaacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.