missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Glucose pentaacetate, 99%

Product Code. 11429657
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429657 50 g 50g
11439657 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429657 Supplier Thermo Scientific Alfa Aesar Supplier No. B22137.18
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 604-68-2 | C16H22O11 | 390.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-68-2
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064071,MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
PubChem CID 2723636
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 109°C to 113°C
Quantity 50 g
Beilstein 98852
Optical Rotation +102° (c=5 in Chloroform)
Formula Weight 390.34
Percent Purity 99%
Chemical Name or Material alpha-D-Glucose pentaacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.