missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Lactose, monohydrate, ACS reagent, ACROS Organics™

Product Code. 10494691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic bucket
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10494691 500 g Plastic bottle 500g
10712212 2 kg Plastic bottle 2kg
10238433 10 kg Plastic bucket 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10494691 Supplier Thermo Scientific Acros Supplier No. 412975000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5989-81-1 , 63-42-3
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-KPKNDVKVSA-N
Synonym alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x
PubChem CID 104938
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Color White
Ignition Residue 0.03% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 500 g
Beilstein 31, 408
Merck Index 14, 5343
Specific Rotation Condition + 54.40 (20.00°C c=10,H2O,NH3)
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 360.3
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material α-D-Lactose, monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.