missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Lactose monohydrate

Product Code. 11448933
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448933 5000 g 5000g
11438933 250 g 250g
11428933 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448933 Supplier Thermo Scientific Alfa Aesar Supplier No. A11074.0I
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol

alpha-D-Lactose monohydrate is used as a carrier and stabiliser of aromas, pharmaceutical products, Food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5989-81-1
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-KPKNDVKVSA-N
Synonym alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x
PubChem CID 104938
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Melting Point 207°C to 211°C (decomposition)
Density 1.525
Odor Odorless
Quantity 5000 g
Beilstein 3768231
Merck Index 14,5343
Solubility Information Soluble in water.
Optical Rotation +52° (c=4.5 in water, 22h)
Formula Weight 360.32 (342.30 Anhydrous)
Chemical Name or Material alpha-D-Lactose monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.