missing translation for 'onlineSavingsMsg'
Learn More

alpha-Fluorophenylacetic acid, 97%

Product Code. 11341397
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
Unit Size:
100mg
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341397 500 mg 500mg
11361397 100 mg 100mg
11351397 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11341397 Supplier Thermo Scientific Alfa Aesar Supplier No. L01745.ME
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1578-63-8 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1578-63-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004221
InChI Key ATPPNMLQNZHDOG-UHFFFAOYSA-N
Synonym alpha-fluorophenylacetic acid, fluorophenylacetic acid, fluoro phenyl acetic acid, .alpha.-fluorophenylacetic acid, benzeneacetic acid, .alpha.-fluoro, fluorophenyl acetic acid, a-fluorophenylacetic acid, a-fluorobenzeneacetic acid, a-fluorophenyl acetic acid, acmc-1bt6i
PubChem CID 102649
IUPAC Name 2-fluoro-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)F

Specifications

Melting Point 78°C to 82°C
Quantity 500 mg
UN Number UN2811
Beilstein 2087378
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material alpha-Fluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.