missing translation for 'onlineSavingsMsg'
Learn More

Neville and More™ Alpha Glass Bottle

Product Code. 15393408 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inner:
3.3 cm
3.86 cm
4.4 cm
6.2 cm
Color:
Amber
Clear
Unit Size:
Pack of 110
Pack of 127
Pack of 130
Pack of 60
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Color Unit Size
15393408 3.86 cm Amber Pack of 127
15343408 3.3 cm Amber Pack of 110
15313418 4.4 cm Amber Pack of 130
15383408 4.4 cm Clear Pack of 130
15323418 6.2 cm Amber Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15393408
  • ONLINE EXCLUSIVE PRICE
    39.10 €
    27.58 € / Pack of 127
    Save 11.52 €
    29% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

Used to store medical. chemical reagents.

  • Standard round glass bottles
  • Lightweight glass bottles
  • Has same neck, one cap for the entire range
  • Manufactured in clean room
  • Packed in clean shrink wrapped packs for enhanced cleanliness
  • Pilfer proof glass neck

Specifications

Color Amber
Diameter (Metric) Inner 3.86 cm
Material Glass (Type 3)
No. per Case 127
Volume (Metric) Brim 67.5 mL
Closure Included No
Height (Metric) 9.4 cm
Closure Size 2.8 cm
Shape Round
Product Type Alpha Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.