missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Hydroxy Olopatadine (>90%), TRC

Product Code. 30137414
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137414 1 mg 1mg
30008639 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137414 Supplier Toronto Research Chemicals Supplier No. TRCH9487711MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1331822-32-2
Molecular Formula C21 H23 N O4
Molecular Weight (g/mol) 353.41
Synonym 11-[3-(Dimethylamino)propylidene]-6,11-dihydro-α-hydroxydibenz[b,e]oxepin-2-acetic Acid, α-Hydroxyolopatadine
IUPAC Name 2-[11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]-2-hydroxyacetic acid
SMILES CN(C)CCC=C1c2ccccc2COc3ccc(cc13)C(O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H23NO4/c1-22(2)11-5-8-17-16-7-4-3-6-15(16)13-26-19-10-9-14(12-18(17)19)20(23)21(24)25/h3-4,6-10,12,20,23H,5,11,13H2,1-2H3,(H,24,25)
Formula Weight 353.1627
Chemical Name or Material alpha-hydroxy Olopatadine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.