missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hydroxyacetophenone, 97%

Product Code. 10187890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10187890 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10187890 Supplier Thermo Scientific Acros Supplier No. 411900250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 582-24-1 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 582-24-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 83°C to 88°C
Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H8O2
MDL Number MFCD00041829
Quantity 25 g
Synonym 2-hydroxyacetophenone, benzoylcarbinol, glycolophenone, phenacyl alcohol, alpha-hydroxyacetophenone, 2-hydroxy-1-phenylethan-1-one, ethanone, 2-hydroxy-1-phenyl, methanol, benzoyl, omega-hydroxyacetophenone, acetophenone, 2-hydroxy
InChI Key ZWVHTXAYIKBMEE-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)CO
IUPAC Name 2-hydroxy-1-phenylethanone
Molecular Weight (g/mol) 136.15
PubChem CID 68490
ChEBI CHEBI:28341
Formula Weight 136.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material α-Hydroxyacetophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • a-Hydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.