missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Ketoglutaric Acid Monosodium Salt, TRC

Product Code. 30407750
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407750 25 g
30393994 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407750 Supplier Toronto Research Chemicals Supplier No. TRCK19120825G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22202-68-2
Molecular Formula C5H5NaO5
Molecular Weight (g/mol) 168.08
Synonym 2-Oxopentanedioic Acid Monosodium Salt, Monosodium 2-Oxoglutarate, Sodium Hydrogen alpha-Ketoglutarate
IUPAC Name sodium;5-hydroxy-2,5-dioxopentanoate
SMILES [Na+].OC(=O)CCC(=O)C(=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H6O5.Na/c6-3(5(9)10)1-2-4(7)8;/h1-2H2,(H,7,8)(H,9,10);/q;+1/p-1
Formula Weight 168.0035
Chemical Name or Material alpha-Ketoglutaric Acid Monosodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.