missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methacrylic Acid, TRC

Product Code. 30004267
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004267 10 g 10g
30005379 5 g 5g
30003335 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004267 Supplier Toronto Research Chemicals Supplier No. TRCM22594510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-41-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
Synonym 2-Methyl-2-propenoic Acid, Methacrylic Acid, 2-Methyl-2-propenoate, 2-Methyl-2-propenoic Acid, 2-Methylacrylic Acid, Acryester MAA, GE 100, GE 110, Isobutenic Acid, Light Ester A, Loctite 3298, MAA, Methylacrylic Acid, NSC 7393, Norsocryl MAA, α-Methylacrylic Acid
IUPAC Name 2-methylprop-2-enoic acid
SMILES C=C(C(O)=O)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H6O2/c1-3(2)4(5)6/h1H2,2H3,(H,5,6)
Formula Weight 86.0368
Percent Purity >95%
Chemical Name or Material alpha-Methacrylic Acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.