missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-2-naphthalenemethanol, 99%, Thermo Scientific™

Product Code. 11934661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11934661 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11934661 Supplier Thermo Scientific Acros Supplier No. 440740050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7228-47-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 74°C to 76°C
Color White to Yellow
Boiling Point 140°C to 142°C (4.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C12H12O
Quantity 5 g
Synonym 1-2-naphthyl ethanol, 1-naphthalen-2-yl ethanol, +/--1-2-naphthyl ethanol, 1-naphthalen-2-yl ethan-1-ol, alpha-methylnaphthalene-2-methanol, .alpha.-methyl-2-naphthalenemethanol, pubchem9427, acmc-209gxz, acmc-1azul, methyl 2-naphtylcarbinol
InChI Key AXRKCRWZRKETCK-UHFFFAOYSA-N
SMILES CC(C1=CC2=CC=CC=C2C=C1)O
IUPAC Name 1-naphthalen-2-ylethanol
Molecular Weight (g/mol) 172.23
PubChem CID 98243
Formula Weight 172.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material α-Methyl-2-naphthalenemethanol
Show More Show Less
Product Identifier
  • alpha-Methyl-2-naphthalenemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.