missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-DL-phenylalanine, 98%, Thermo Scientific™

Product Code. 11456237
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456237 0.25 g 0.25g
11436237 1 g 1g
11446237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456237 Supplier Thermo Scientific Alfa Aesar Supplier No. B25155.01
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-26-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00010512
InChI Key IRZQDMYEJPNDEN-UHFFFAOYNA-N
Synonym alpha-methyl-dl-phenylalanine, 2-amino-2-methyl-3-phenylpropionic acid, alpha-methylphenylalanine, h-dl-me phe-oh, h-alpha-me-dl-phe-oh, +/--2-amino-2-methyl-3-phenylpropionic acid, phenylalanine, alpha-methyl, 2-amino-2-methyl-3-phenyl-propanoic acid, h-me-dl-phe-oh, 2-methylphenylalanine #
PubChem CID 108055
IUPAC Name 2-amino-2-methyl-3-phenylpropanoic acid
SMILES CC(C(N)C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 293°C to 294°C (decomposition)
Quantity 0.25 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material alpha-Methyl-DL-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.