missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(Methylenecyclopropyl)glycine (Mixture of Diastereomers), TRC

Product Code. 30007733
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007733 1 mg 1mg
30003466 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007733 Supplier Toronto Research Chemicals Supplier No. TRCM3031501MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2517-07-9
Molecular Formula C6 H9 N O2
Molecular Weight (g/mol) 127.14
Synonym α-Amino-2-methylenecyclopropaneacetic Acid
IUPAC Name 2-amino-2-(2-methylidenecyclopropyl)acetic acid
SMILES NC(C1CC1=C)C(=O)O

Specifications

InChI Formula InChI=1S/C6H9NO2/c1-3-2-4(3)5(7)6(8)9/h4-5H,1-2,7H2,(H,8,9)
Quantity 1 mg
Formula Weight 127.0633
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material rac-alpha-(Methylenecyclopropyl)glycine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.