missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylglyceric Acid, TRC

Product Code. 30134754
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134754 25 g 25g
30132555 2.5 g 2.5g
30011456 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134754 Supplier Toronto Research Chemicals Supplier No. TRCM31150525G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21620-60-0
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.1
Synonym 2,3-Dihydroxy-2-methylpropanoic Acid, 2,3-Dihydroxy-2-methylpropionic Acid, 2-C-Methylglyceric Acid, 2-Methyl-2,3-dihydroxypropionic Acid, 2-Methylglyceric Acid, 2-Methylglyceronic Acid, α,β-Dihydroxyisobutyric Acid
SMILES Cl.OCC1C(O)C(O)C(N)C(O)O1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H8O4/c1-4(8,2-5)3(6)7/h5,8H,2H2,1H3,(H,6,7)
Chemical Name or Material alpha-Methylglyceric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.