missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylphenylacetonitrile, 96%

Product Code. 11450214
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450214 10 g 10g
11334609 25 g 25g
11460214 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11450214 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A13636.09
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format
Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1823-91-2 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1823-91-2
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001871
InChI Key NVAOLENBKNECGF-UHFFFAOYNA-N
Synonym 2-phenylpropionitrile, hydratroponitrile, alpha-methylphenylacetonitrile, alpha-methylbenzyl cyanide, 2-phenylpropiononitrile, benzeneacetonitrile, .alpha.-methyl, methylbenzyl cyanide, .alpha.-phenylpropionitrile, .alpha.-phenylethyl cyanide, .alpha.-methylbenzyl cyanide
PubChem CID 15761
IUPAC Name 2-phenylpropanenitrile
SMILES CC(C#N)C1=CC=CC=C1

Especificaciones

Density 0.985
Boiling Point 230°C to 232°C
Flash Point 93°C (199°F)
Refractive Index 1.5106
Quantity 10 g
UN Number UN3276
Beilstein 774576
Formula Weight 131.18
Percent Purity 97%
Chemical Name or Material alpha-Methylphenylacetonitrile
compliance-icons
Nombre del producto
  • alpha-Methylphenylacetonitrile
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Otros peligros
  • MIXTURE LIST-Contain: alpha-Methylphenylacetonitrile

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.