missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Methylstyrene, TRC

Product Code. 30482916
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482916 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482916 Supplier Toronto Research Chemicals Supplier No. TRCM32539550ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Methylstyrene
CAS 98-83-9
Molecular Formula C9 H10
Quantity 50 mL
Synonym Benzene, isopropenyl- (7CI), Styrene, α-methyl- (8CI), (1-Methylethenyl)benzene, 1-Methyl-1-phenylethene, 1-Methyl-1-phenylethylene, 1-Methylvinyl benzene, 1-Phenyl-1-methylethylene, 1-Propene, 2-phenyl-, 2-Phenyl-1-propene, 2-Phenyl-1-propylene, 2-Phenyl-2-propene, 2-Phenylpropene, 2-Phenylpropylene, Isopropenylbenzene, K 150 (styrene), Methylstyrene, NSC 9400, Prop-1-en-2-ylbenzene, α-Methylstyrene, α-Methylstyrol, α-Methylvinylbenzene, β-Phenylpropylene
InChI Formula InChI=1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3
SMILES CC(=C)c1ccccc1
IUPAC Name prop-1-en-2-ylbenzene
Molecular Weight (g/mol) 118.18
Formula Weight 118.0783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.