missing translation for 'onlineSavingsMsg'
Learn More

alpha-Naphthoflavone, 97%

Product Code. 10794071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10794071 5 g Glass bottle 5g
10753891 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10794071 Supplier Thermo Scientific Acros Supplier No. 128160050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 604-59-1 | C19H12O2 | 272.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-59-1
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.29
MDL Number MFCD00004985
InChI Key VFMMPHCGEFXGIP-UHFFFAOYSA-N
Synonym alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one
PubChem CID 11790
ChEBI CHEBI:76995
IUPAC Name 2-phenylbenzo[h]chromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 155°C to 158°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, IV, 5551
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,chloroform,soluble in sulfuric acid
Formula Weight 272.29
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α-Naphthoflavone, 99.5+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.