missing translation for 'onlineSavingsMsg'
Learn More

alpha-Naphthol Orange (Technical Grade), TRC

Product Code. 30478070
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478070 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478070 Supplier Toronto Research Chemicals Supplier No. TRCN369040250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Orange 1 sodium salt
CAS 523-44-4
Molecular Formula C16 H11 N2 O4 S . Na
Quantity 250 mg
Synonym Benzenesulfonic acid','4-[(4-hydroxy-1-naphthalenyl)azo]-','monosodium salt (9CI), C.I. Acid Orange 20 (7CI), C.I. Acid Orange 20','monosodium salt (8CI), 1333 Orange, 4-p-Sulfophenylazo-1-naphthol monosodium salt, A.F. Orange No. 1, Acid Leather Orange I, Acid Orange 20, Acid Orange I, Aizen Orange I, C.I. 14600, Certiqual Orange I, Dye orange No. 1, Eniacid Orange I, Ext D & C Orange 3, Ext D and C Orange No. 3, External D and C Orange Number 3, FD and C Orange 1, FD and C Orange No. 1, Hispacid Orange 1, Japan Orange 1, Japan Orange 402, Japan Orange No. 402, Java Orange I, Naphthalene Orange I, Naphthol Orange, Neklacid Orange 1, Orange 1 sodium salt, Orange 402, Orange I, Orange I Extra Conc. A Export, Orange IM, Orange No. 402, Tertracid Orange I, Tropaeolin 1, Tropaeolin OOO 1, Yellow 404, α-Naphthol Orange
InChI Formula InChI=1S/C16H12N2O4S.Na/c19-16-10-9-15(13-3-1-2-4-14(13)16)18-17-11-5-7-12(8-6-11)23(20,21)22;/h1-10,19H,(H,20,21,22);/q;+1/p-1
SMILES [Na+].Oc1ccc(N=Nc2ccc(cc2)S(=O)(=O)[O-])c3ccccc13
IUPAC Name sodium;4-[(E)-(4-hydroxynaphthalen-1-yl)diazenyl]benzenesulfonate
Molecular Weight (g/mol) 350.32
Formula Weight 350.0337
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.