missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Naphthol Orange, TRC
Specifications
Specifications
| Chemical Name or Material | Orange 1 sodium salt |
| CAS | 523-44-4 |
| Molecular Formula | C16 H11 N2 O4 S . Na |
| Quantity | 500 mg |
| Synonym | Benzenesulfonic acid','4-[(4-hydroxy-1-naphthalenyl)azo]-','monosodium salt (9CI), C.I. Acid Orange 20 (7CI), C.I. Acid Orange 20','monosodium salt (8CI), 1333 Orange, 4-p-Sulfophenylazo-1-naphthol monosodium salt, A.F. Orange No. 1, Acid Leather Orange I, Acid Orange 20, Acid Orange I, Aizen Orange I, C.I. 14600, Certiqual Orange I, Dye orange No. 1, Eniacid Orange I, Ext D & C Orange 3, Ext D and C Orange No. 3, External D and C Orange Number 3, FD and C Orange 1, FD and C Orange No. 1, Hispacid Orange 1, Japan Orange 1, Japan Orange 402, Japan Orange No. 402, Java Orange I, Naphthalene Orange I, Naphthol Orange, Neklacid Orange 1, Orange 1 sodium salt, Orange 402, Orange I, Orange I Extra Conc. A Export, Orange IM, Orange No. 402, Tertracid Orange I, Tropaeolin 1, Tropaeolin OOO 1, Yellow 404, α-Naphthol Orange |
| InChI Formula | InChI=1S/C16H12N2O4S.Na/c19-16-10-9-15(13-3-1-2-4-14(13)16)18-17-11-5-7-12(8-6-11)23(20,21)22;/h1-10,19H,(H,20,21,22);/q;+1/p-1 |
| SMILES | [Na+].Oc1ccc(N=Nc2ccc(cc2)S(=O)(=O)[O-])c3ccccc13 |
| IUPAC Name | sodium;4-[(E)-(4-hydroxynaphthalen-1-yl)diazenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 350.32 |
| Formula Weight | 350.0337 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?