missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Naphthylflavone, TRC

Product Code. 17939084
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17939084 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17939084 Supplier Toronto Research Chemicals Supplier No. TRCN3725051G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 604-59-1
Molecular Formula C19 H12 O2
Quantity 1 g
InChI Formula InChI=1S/C19H12O2/c20-17-12-18(14-7-2-1-3-8-14)21-19-15-9-5-4-6-13(15)10-11-16(17)19/h1-12H
SMILES O=C1C=C(Oc2c1ccc3ccccc23)c4ccccc4
IUPAC Name 2-phenylbenzo[h]chromen-4-one
Molecular Weight (g/mol) 272.3
Formula Weight 272.0837
Chemical Name or Material alpha-Naphthoflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.