missing translation for 'onlineSavingsMsg'
Learn More

(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer

Product Code. 11361697
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361697 50 g 50g
11371697 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361697 Supplier Thermo Scientific Alfa Aesar Supplier No. L04759.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7785-26-4 | C10H16 | 136.238 g/mol

(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer, is used in food and beverage (incl. additives, contaminants), fragrance / essential oil, nutraceutical. It is also used as a anti- infective agent. It is used as a component of pine resin and a useful chiral synthetic intermediate. Alpha-pinene is the most widely encountered terpenoid in nature and is highly repellant to insects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer, is used in food and beverage (incl. additives, contaminants), fragrance / essential oil, nutraceutical. It is also used as a anti- infective agent. It is used as a component of pine resin and a useful chiral synthetic intermediate. Alpha-pinene is the most widely encountered terpenoid in nature and is highly repellant to insects.

Solubility
Soluble in chloroform, ether, alcohol, and methanol. Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-26-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00064145
InChI Key GRWFGVWFFZKLTI-IUCAKERBSA-N
Synonym --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s
PubChem CID 440968
ChEBI CHEBI:28660
IUPAC Name (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CCC2CC1C2(C)C

Specifications

Melting Point -64°C
Boiling Point 154°C to 156°C
Flash Point 32°C (89°F)
Quantity 50 g
UN Number UN2368
Beilstein 1903790
Merck Index 14,7445
Optical Rotation −42° (Neat)
Formula Weight 136.24
Percent Purity 98%
Assay cont. variable amounts of enantiomer
Chemical Name or Material (-)-alpha-Pinene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.