missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol, 96%

Product Code. 11387168
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387168 500 g 500g
15451077 25 g 25g
11397168 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387168 Supplier Thermo Scientific Alfa Aesar Supplier No. 016285.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

A naturally-occuring monoterpene alcohol | CAS: 98-55-5 | C10H18O | 154.253 g/mol

A naturally-occuring monoterpene alcohol.Alpha-terpineol is used as an antioxidant, antiseptic, antihypernociception and anti-inflammatory. It is also used as a solvent. It is an important ingredient of pine oil disinfectants. Further, it is used as a fragrance in perfumes, fat denaturant for soap production and synthetic flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Evaporation Residue 0.002%, Hydrogen sulfide (H2S) P.T. (limit about 1.5ppm, Sulfur dioxide (SO2) P.T. (limit about 2.5ppm), H2O 0.05%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-55-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001557
InChI Key WUOACPNHFRMFPN-UHFFFAOYSA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O

Specifications

Melting Point 31°C to 32°C
Color Colorless to Yellow
Density 0.930 g/mL
Boiling Point 210°C to 219°C
Flash Point 90°C (194°F)
Refractive Index 1.483
Quantity 500 g
Merck Index 14,9171
Solubility Information Miscible with water,benzene,propylene glycol,acetone,alcohol and ether.
Formula Weight 154.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material alpha-Terpineol
Show More Show Less
compliance-icons
Product Identifier
  • alpha-Terpineol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: .alpha.-Terpineol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.