missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (±)-alpha-Tocopherol, 95%, synthetic

Product Code. 10077789 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10077789 100 g Glass bottle 100g
10785242 25 g Glass bottle 25g
10482770 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10077789 Supplier Thermo Scientific Chemicals Supplier No. 428121000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 2.5°C to 3.5°C
Density 0.9500g/mL
Boiling Point 393.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 9653
Specific Gravity 0.95
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in oils, fats, acetone, alcohol,, chloroform, diethyl ether, other fat solvents.
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Viscosity 4200 mPa.s (20°C)
Formula Weight 430.71
Percent Purity 95%
Chemical Name or Material (±)-α-Tocopherol, Synthetic
Show More Show Less
compliance-icons
Product Identifier
  • (+/-)-alpha-Tocopherol
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.