missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Toluenesulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Alpha-Toluenesulfonyl Chloride |
| CAS | 1939-99-7 |
| Molecular Formula | C7H7ClO2S |
| Quantity | 5 g |
| Synonym | Benzylsulfonyl Chloride, NSC 41894, Phenylmethanesulfonyl Chloride |
| InChI Formula | InChI=1S/C7H7ClO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2 |
| SMILES | ClS(CC1=CC=CC=C1)(=O)=O |
| IUPAC Name | phenylmethanesulfonyl chloride |
| Molecular Weight (g/mol) | 190.65 |
| Formula Weight | 189.99 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?