missing translation for 'onlineSavingsMsg'
Learn More

alpha-Toluenesulfonyl fluoride, 99%

Product Code. 11479667
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11479667

Brand: Thermo Scientific Alfa Aesar B22146.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 329-98-6 | C7H7FO2S | 174.189 g/mol

α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

Solubility
Soluble in ether, benzene, anhydrous ethanol, methanol, 2- propane. Hydrolyses in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, amines, bases, water/ moisture. Store at 4°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00007424
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

Melting Point 92°C to 94°C
Quantity 5 g
UN Number UN2923
Beilstein 2088311
Sensitivity Moisture sensitive
Solubility Information Soluble in ether,benzene,anhydrous ethanol,methanol,2- propane. Hydrolyses in water.
Formula Weight 174.19
Percent Purity 99%
Chemical Name or Material alpha-Toluenesulfonyl fluoride
compliance-icons compliance-icons
Identificativo prodotto
  • alpha-Toluenesulfonyl fluoride
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicazioni di pericolo
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contains : Benzenemethanesulfonyl fluoride

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato