missing translation for 'onlineSavingsMsg'
Learn More

Alphacel Hydrolyzed, MP Biomedicals™

Product Code. 11471392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 lb.
Unit Size:
5lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471392 5 lb. 5lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471392 Supplier MP Biomedicals Supplier No. 0290045405
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Alphacel Hydrolyzed, White Powder

  • Alphacel is a polysaccharide composed of long chains of β(1,4) linked D-glucose units.
  • Cellulose is used in a wide range of applications that require a structural back-bone.
  • It is used in the preparation of chromatographic structural media, in gels for processes such as electrophoresis and ionic separations and as a substrate to identify and study cellulosic systems
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
MDL Number MFCD00081512
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N

Specifications

Color White
pH 5.5 to 7.0
Physical Form Powder
Quantity 5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.