missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ Alphachem X350 Disposable Coverall

Product Code. 31702826
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31702826 Medium Pack of 10
31702838 Large Pack of 10
31702820 X-Large Pack of 10
31702842 2X-Large Pack of 10
31702837 3X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31702826 Supplier Skytec™ Supplier No. ASC00002GF
  • ONLINE EXCLUSIVE PRICE
    1114.13 €
    713.80 € / Pack of 10
    Save 400.33 €
    35% Off
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

X350 disposable coveralls provide a high barrier against chemicals and infective agents.

  • Manufactured using polyethylene and EVOH barrier composite, bonded structural spun-bonded and polypropylene base layer suitable for higher-risk applications
  • Tough and durable
  • Improved fit, enhanced freedom of movement with a reinforced crotch
  • Double zip system and grab tag to support with an emergency
  • Easy to hold when wearing gloves
  • Reinforced knee patches for extra durability when kneeling
  • Stitched and fully taped seam stitching and elastication
  • Elasticated wrists and ankles for secure fit
  • Three-piece hood construction for better fit
  • Available in a wide range of sizes
  • Double sleeve system with inner thumb loop ensuring arm protection to support when arm is raised

Specifications

Product Type Chemical-resistant Coveralls
Size Medium
Gender Unisex
Material Composite Fabric
Color Orange
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals, Viruses
Certifications/Compliance CE III or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5 or - EN 14126:2003 + AC:2004 Type 3B,Type 4B,Type 5B,Type 6B or - EN 14605:2005 + A1:2009 Type 3,Type 4
For Use With (Application) Chemical Handling and Containment, Commercial Washdown, Transportation of Fuels
Weight (Metric) 731 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.