missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ AlphaGuard™ Frocks

Product Code. 15277198
Change view
Click to view available options
Size:
X-Large
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15277198 X-Large Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15277198 Supplier AlphaProTech Supplier No. FK321224

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Frocks are constructed with low particle count multilayer SMS material to offer high levels of breathability without compromising filtration efficiency.

  • Low particle count
  • Fluid repellent
  • Clean, durable and soft
  • Static dissipative
  • Raglan sleeves
  • Snap collar and close
  • Serged seams

Specifications

Size X-Large
Material SMS
Color White
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Mandarin
Size (English) Chest 24.75 in.
Size (Metric) Chest 62.8 cm
Sleeve Style Raglan Sleeve
Length (Metric) Sleeve 92.71 cm
Product Type Controlled Environments Frock
Seam Type Serged
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.